
TSKgel® QC-PAK GFC 300 HPLC Column

Synonym(s):
TSKgel® SWXL Columns
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing