
Sciadopitysin

Synonym(s):
4′,4′′′,7-Trimethylamentoflavone, 5,7-Dihydroxy-8-(5-(5-hydroxy-7-methoxy-4-oxo-4H-1-benzopyran-2-yl)-2-methoxyphenyl)-2-(4-methoxyphenyl)-4-benzopyrone, Amentoflavone-7,4′,4”′- trimethylether
Empirical Formula (Hill Notation):
C33H24O10
CAS Number:
Molecular Weight:
580.54
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing