
Riboflavin for peak identification

Synonym(s):
(−)-Riboflavin, Lactoflavin, Vitamin B2, Vitamin G
Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Products
Product Category
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing