
Mepivacaine impurity B

Synonym(s):
Desbutylbupivacaine, (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide
Empirical Formula (Hill Notation):
C14H20N2O
CAS Number:
Molecular Weight:
232.32
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing