
L-Cysteine S-sulfate

Synonym(s):
S-Sulfo-L-cysteine
Empirical Formula (Hill Notation):
C3H7NO5S2
CAS Number:
Molecular Weight:
201.22
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing