
Lauryl alcohol

Synonym(s):
1-Dodecanol, Alcohol C12, Dodecyl alcohol, Lauryl alcohol
Linear Formula:
CH3(CH2)11OH
CAS Number:
Molecular Weight:
186.33
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3