
Iprovalicarb

Synonym(s):
Nα-(Isopropoxycarbonyl)-N-[1-(p-tolyl)ethyl]-L-valinamide, Isopropyl (S)-2-methyl-1-[1-(p-tolyl)ethylcarbamoyl]propylcarbamate
Empirical Formula (Hill Notation):
C18H28N2O3
CAS Number:
Molecular Weight:
320.43
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2