
Hydroxocobalamin chloride

Synonym(s):
Vitamin B12a hydrochloride, Hydroxocobalamin hydrochloride
Empirical Formula (Hill Notation):
C62H89CoN13O15P · HCl
CAS Number:
Molecular Weight:
1382.82
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing