Hexacosanol
Synonym(s):
1-Hexacosanol, Ceryl alcohol, Hexacosyl alcohol
Linear Formula:
CH3(CH2)25OH
CAS Number:
Molecular Weight:
382.71
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing