Skip to Content
Merck
(Ethylenediamine)palladium(II) chloride ≥99.99%

(Ethylenediamine)palladium(II) chloride

(Ethylenediamine)palladium(II) chloride ≥99.99%
Synonym(s):
Dichloro(1,2-diaminoethane)palladium, Dichloro(ethylenediamine)palladium(II), Palladium(II) chloride ethylenediamine complex
Linear Formula:
Pd(H2NCH2CH2NH2)Cl2
CAS Number:
Molecular Weight:
237.42

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing