
Doxazosin Related Compound E

Synonym(s):
2,4-Dichloro-6,7-dimethoxyquinazoline
Empirical Formula (Hill Notation):
C10H8Cl2N2O2
CAS Number:
Molecular Weight:
259.09
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2