
Desoxycorticosterone pivalate

Synonym(s):
Percorten pivalate
Empirical Formula (Hill Notation):
C26H38O4
CAS Number:
Molecular Weight:
414.58
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing