Skip to Content
Merck
Decyl β-D-maltopyranoside ≥98% (GC)

Decyl β-D-maltopyranoside

Decyl β-D-maltopyranoside ≥98% (GC)
Synonym(s):
Decyl-β-D-maltoside
Empirical Formula (Hill Notation):
C22H42O11
CAS Number:
Molecular Weight:
482.56

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing