![[D-Ala6]-LH-RH acetate salt hydrate ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/222/268/0818f701-a831-440e-9dd2-5f851035c421/640/0818f701-a831-440e-9dd2-5f851035c421.png)
[D-Ala6]-LH-RH acetate salt hydrate
![[D-Ala6]-LH-RH acetate salt hydrate ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/222/268/0818f701-a831-440e-9dd2-5f851035c421/640/0818f701-a831-440e-9dd2-5f851035c421.png)
Synonym(s):
(D-Ala-6) Luteinizing Hormone Releasing Hormone
Empirical Formula (Hill Notation):
C56H77N17O13 · xC2H4O2 · yH2O
Molecular Weight:
1196.32 (anhydrous free base basis)
Products
Product Category
Brand
Formula Weight
Gene Alias
Purity
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing