
Cromolyn Related Compound A

Synonym(s):
1,3-Bis(2-acetyl-3-hydroxyphenoxy)propan-2-ol
Empirical Formula (Hill Notation):
C19H20O7
CAS Number:
Molecular Weight:
360.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2