
Chlorthiophos

Synonym(s):
O-(2,5-Dichloro-4-(methylthio)phenyl) O,O-diethyl phosphorothioate (predominant isomer)
Empirical Formula (Hill Notation):
C11H15Cl2O3PS2
CAS Number:
Molecular Weight:
361.24
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing