
Carbimazole

Synonym(s):
1-Ethoxycarbonyl-3-methyl-2-thio-4-imidazoline, 1-Methyl-3-carbethoxy-2-thioglyoxalone, 2,3-Dihydro-3-methyl-2-thioxo-1H-imidazole-1-carboxylic acid ethyl ester, Athyromazole, Carbethoxymethimazole, Carbimazol, Ethyl 3-methyl-2-thioimidazoline-1-carboxylate, Carbimazole, 2,3-Dihydro-3-methyl-2-thioxo-1H-imidazole-1-carboxylic acid ethyl ester
Empirical Formula (Hill Notation):
C7H10N2O2S
CAS Number:
Molecular Weight:
186.23
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique