
Bupropion Hydrochloride Related Compound F

Synonym(s):
1-(3-Chlorophenyl)-1-hydroxy-2-propanone
Empirical Formula (Hill Notation):
C9H9ClO2
CAS Number:
Molecular Weight:
184.62
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing