
Benzovindiflupyr

Synonym(s):
N-[9-(Dichloromethylene)-1,2,3,4-tetrahydro-1,4-methanonaphthalen-5-yl]-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C18H15Cl2F2N3O
CAS Number:
Molecular Weight:
398.23
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing