
Anti-CALD1 antibody produced in rabbit

Synonym(s):
Anti-CDM antibody produced in rabbit, Anti-Caldesmon antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing