
4-Hydroxytamoxifen

Synonym(s):
4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol, 4-OHT, cis/trans-4-Hydroxytamoxifen, cis/trans-4-Hydroxytamoxifen
Empirical Formula (Hill Notation):
C26H29NO2
CAS Number:
Molecular Weight:
387.51
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Format
Research Area
Quality Segment
Application
Mode of Action
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing