
3-(Trimethoxysilyl)propyl acrylate

Synonym(s):
(3-Acryloyloxypropyl)trimethoxysilane
Linear Formula:
H2C=CHCO2(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
234.32
Products
Product Category
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing