
2-Aminoflubendazole

Synonym(s):
(2-Amino-1H-benzoimidazol-5-yl)-(4-fluoro-phenyl)-methanone, 2-Amino-5-(4-fluorobenzoyl)-1H-benzimidazole, Hydrolyzed flubendazole
Empirical Formula (Hill Notation):
C14H10FN3O
CAS Number:
Molecular Weight:
255.25
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing