Skip to Content
Merck
2′,4′-Dihydroxy-3′-propylacetophenone analytical standard

2′,4′-Dihydroxy-3′-propylacetophenone

2′,4′-Dihydroxy-3′-propylacetophenone analytical standard
Linear Formula:
CH3CH2CH2C6H2(OH)2COCH3
CAS Number:
Molecular Weight:
194.23

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2