Skip to Content
Merck
1,7-Dimethyluric acid ≥97.0% (HPLC)

1,7-Dimethyluric acid

1,7-Dimethyluric acid ≥97.0% (HPLC)
Synonym(s):
1,7-Dimethyl-2,6,8-trihydroxypurine
Empirical Formula (Hill Notation):
C7H8N4O3
CAS Number:
Molecular Weight:
196.16
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing