Skip to Content
Merck
All Photos(1)

Key Documents

78823

Supelco

Xanthurenic acid

analytical standard

Synonym(s):

4,8-Dihydroxyquinaldic acid, 4,8-Dihydroxyquinoline-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO4
CAS Number:
Molecular Weight:
205.17
Beilstein:
185954
MDL number:
UNSPSC Code:
12352100

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

form

solid

shelf life

limited shelf life, expiry date on the label

mp

297-298 °C (dec.) (lit.)

application(s)

clinical
clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)c1cc(O)c2cccc(O)c2n1

InChI

1S/C10H7NO4/c12-7-3-1-2-5-8(13)4-6(10(14)15)11-9(5)7/h1-4,12H,(H,11,13)(H,14,15)

InChI key

FBZONXHGGPHHIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service