Skip to Content
Merck
All Photos(1)

Key Documents

NIST1869

Infant/Adult Nutritional Formula II (milk/whey/soy-based)

NIST® SRM® 1869, pkg of (10 pouches x 10 g each), storage temp.:-20°C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

manufacturer/tradename

NIST® 1869

packaging

pkg of (10 pouches x 10 g each)

format

matrix material

shipped in

dry ice

storage temp.

−20°C

General description

This SRM is intended primarily for validation of methods for determining proximates, fatty acids, cholesterol, vitamins, elements, amino acids, and nucleotides in infant and adult nutritional formulas and similar materials. This SRM can also be used for quality assurance when assigning values to in-house reference materials. The SRM is a hydrolyzed protein based, hybrid infant/adult nutritional powder prepared by a manufacturer of infant formula and adult nutritional products.A unit of SRM 1869 consists of ten packets, each containing approximately 10 g of material.Additional Notes: SRM 1869 is intended for research use, NOT FOR HUMAN CONSUMPTION. This material contains some nutrients at levels not permitted in infant formula and is NOT AN INFANT FORMULA. See Certificate of Analysis for storage and use instructions.

SRM 1869_cert

SRM 1869_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service