Skip to Content
Merck
All Photos(1)

Key Documents

ERMAC626

Arsenobetaine in water

ERM®, certified reference material

Synonym(s):

Arsenobetaine solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
3933180
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

SMILES string

C[As+](C)(C)CC([O-])=O

InChI

1S/C5H11AsO2/c1-6(2,3)4-5(7)8/h4H2,1-3H3

InChI key

SPTHHTGLGVZZRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ERM is a registered trademark of European Commission

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service