Skip to Content
Merck
All Photos(1)

Key Documents

RU5500

Sigma-Aldrich

Rabies Detection Reagent - Direct Fluorescence Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

103.7 & 502.2, monoclonal

species reactivity

virus

packaging

antibody small pack of 5 mL

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

2-8°C

General description

The Rabies Detection Reagent is a blend of two rabies virus monoclonal antibodies, clones 103.7and 502.2 (Cat. No. RU5500). Each antibody is conjugated to FITC, blended, and tested for use in Immunofluorescence for the detection of Rabies.

Application

Meets specifications.

This mouse monoclonal antibody blend to Rabies, Cat. No. RU5500 is conjugated to FITC and tested for use in Immunofluorescence for the detection of Rabies.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified in PBS containing 1% BSA, 0.05% Tween-20 and 0.1% sodium azide .

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

1 mg/mL. Titer should be determined for specific sample.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service