Skip to Content
Merck
All Photos(3)

Key Documents

MABE955

Sigma-Aldrich

Anti-phospho RNA Pol II (Ser7), clone 3D4A12 Antibody

clone 3D4A12, 1 mg/mL, from rat

Synonym(s):

DNA-directed RNA polymerase II subunit RPB1, RNA polymerase II subunit B1, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RNA PolII, RNAPIIS7p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3D4A12, monoclonal

species reactivity

canine, human, monkey, rat, mouse

concentration

1 mg/mL

technique(s)

ChIP: suitable (ChIP-seq)
ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer7)

Gene Information

human ... POLR2A(5430)

General description

As the study of the RNA polymerase II (RNAPII) transcription complex expands, many researchers are interested in using immunoblots to detect RNAPII in various protein preparations and column fractions. HeLa cell RNAPII contains 10 subunits, the largest possessing a heptapeptide repeat (PTSPSYS) in the C-terminal domain (CTD). This large subunit cycles, during the course of transcription, through nonphosphorylated (RNAPIIA) and hyperphosphorylated (RNAPIIO) forms. RNAPIIA associates with the basal transcription complex and becomes phosphorylated to the RNAPIIO form before the synthesis of the first phosphodiester bond. The timing of these events suggests that this phosphorylation event may function as a regulatory point, and there is considerable interest in identifying components of the transcription complex with CTD kinase activity.

Immunogen

KLH-conjugated linear peptide corresponding to human phospho RNA Pol II (Ser7).

Application

Chromatin Immunoprecipitation-Sequencing Analysis: A representative lot from an independent laboratory immunoprecipitated phospho RNA Pol II (Ser7) from HeLa cell lysate (Maehara, K., et al. (2013). Nucleic Acids Res. 41(1):54-62.).

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated phospho RNA Pol II (Ser7) from HeLa cell lysate (Maehara, K., et al. (2013). Nucleic Acids Res. 41(1):54-62.).

Western Blotting Analysis: 1 µg/mL from a representative lot detected phospho RNA Pol II (Ser7) in HeLa (lane 1), COS1, L929, NRK, MDCK, and DM4 whole cell extracts (Prof. Taro Tachibana, Cell Engineering Corporation.).

Immunocytochemistry Analysis: A 1:500 dilution from representative lot detected phospho RNA Pol II (Ser7) in HeLa cells (Prof. Taro Tachibana, Cell Engineering Corporation.).

ELISA Analysis: A representative lot detected phospho RNA Pol II (Ser7) in a panel of unmodified and modified RNA Pol II peptides (Prof. Taro Tachibana, Cell Engineering Corporation.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
This Anti-phospho RNA Pol II (Ser7) antibody, clone 3D4A12 is validated for use in western blotting, ChIP-seq, ChIP, ICC & ELISA for the detection of phospho RNA Pol II (Ser7).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected phospho RNA Pol II (Ser7) in 10 µg of HeLa cell lysate.

Target description

~270 kDa observed. Uncharacterized band(s) may be observed in some cell lysates. This protein may be observed at higher molecular weights due to post-translational modification.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service