Skip to Content
Merck
All Photos(1)

Key Documents

MAB5102

Sigma-Aldrich

Mouse IgG1/IgG2a Neg. Control Antibody, clones 131-2G /131-2A Antibody, FITC conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody product type

primary antibodies

species reactivity

all

packaging

vial of 1 mL

manufacturer/tradename

Millipore

technique(s)

immunofluorescence: suitable

shipped in

2-8°C

storage temp.

2-8°C

Features and Benefits

This mouse monoclonal blend Mouse IgG1/IgG2a Negative Control, Cat. No. MAB5102 is tested for use in Immunofluorescence.

Target description

Mouse IgG1/IgG2a Isotype Control is designed with RSV antibodies to produce this negative control antibody blend of IgG1 and IgG2a isotypes.This antibody will cross react with respiratory syncytial virus (RSV).

Physical form

Purified antibody blend supplied in PBS, 1% BSA, and 0.0125% Tween 20 containing 0.1% sodium azide.

Storage and Stability

Store at +2°C to +8°C. It is recommended to store the product undiluted and protected from prolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service