Merck
  • Eotaxin-2 increased toll-like receptor 4 expression in endothelial cells in vitro and exacerbates high-cholesterol diet-induced atherogenesis in vivo.
Eotaxin-2 increased toll-like receptor 4 expression in endothelial cells in vitro and exacerbates high-cholesterol diet-induced atherogenesis in vivo.
American journal of translational research (2017-01-13)
Chien-Sung Tsai, Chun-Yao Huang, Chien-Ho Chen, Yi-Wen Lin, Chun-Ming Shih, Nai-Wen Tsao, Kuang-Hsing Chiang, Chi-Yuan Lee, Hellen Jeng, Feng-Yen Lin
RESUMEN
Eotaxin-2 is a potent chemoattractant. High concentration of eotaxin-2 triggers the inflammation and tumor metastasis. Inhibition of eotaxin-2 may protect experimental atherogenesis although the mechanism is still unclear. Toll-like receptor 4 (TLR4) plays a major role mediating vascular inflammation, which is related to atherogenesis. In the results, we demonstrated that eotaxin-2 potentially impairs the tube formation capacity of human coronary artery endothelial cells (HCAECs). Eotaxin-2 augments the monocytic adhesion in lipopolysaccharides (LPS)-induced HCAECs, and which were reversed by TLR4 siRNA. Thus this study was conducted to investigate whether eotaxin-2 increases TLR4 expression, and then enhances the sensitivity of cells to antigen stimulation in HCAECs, which mediates the increasing of the development of serious atherosclerosis. In fact, we showed that JNK/SAPK, p38 MAPK, and ERK1/2 activation contribute to the transcriptional signaling pathway, JNK/SAPK and p38 MAPK regulate post-transcriptional modification, as well as protein-trafficking pathway in eotaxin-2-treated HCAECs TLR4 expression. RNA binding proteins, such as human antigen R (HuR) and tristetraprolin (TTP) mediate stability of TLR4 mRNA and chaperone, such as PRAT4A (a protein associated with TLR4) regulate trafficking of TLR4 protein might confer eotaxin-2 responsiveness. Eotaxin-2 administration led to a significant elevation of high cholesterol diet-induced atherosclerosis, and of TLR4 expression in B6.129S7-Ldlrtm1Her /J but not Ldlr-/--/-/Tlr4-/- mice. Our results revealed that eotaxin-2 induced overexpression TLR4 via mitogen-activated protein kinases (MAPK) signaling pathways, RNA binding proteins-mediated mRNA stabilization, and PRAT4A-regulated trafficking in HCAECs. These effects may lead to amplification of inflammatory responses contribute to the pathogenesis of cardiovascular disorders.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%