Merck
  • Nuclear α1-antichymotrypsin promotes chromatin condensation and inhibits proliferation of human hepatocellular carcinoma cells.
Nuclear α1-antichymotrypsin promotes chromatin condensation and inhibits proliferation of human hepatocellular carcinoma cells.
Gastroenterology (2013-01-09)
Mónica Santamaria, Ana Pardo-Saganta, Laura Alvarez-Asiain, Marianna Di Scala, Cheng Qian, Jesús Prieto, Matías A Avila
RESUMEN
α1-Antichymotrypsin (α1-ACT), a member of the serpin family (SERPINA3), is an acute-phase protein secreted by hepatocytes in response to cytokines such as oncostatin M. α1-ACT is a protease inhibitor thought to limit tissue damage produced by excessive inflammation-associated proteolysis. However, α1-ACT also is detected in the nuclei of cells, where its activities are unknown. Expression of α1-ACT is down-regulated in human hepatocellular carcinoma (HCC) tissues and cells; we examined its roles in liver regeneration and HCC proliferation. We measured levels of α1-ACT messenger RNA in human HCC samples and healthy liver tissue. We reduced levels of α1-ACT using targeted RNA interference in human HCC (HepG2) and mouse hepatocyte (AML12) cell lines, and overexpressed α1-ACT from lentiviral vectors in Huh7 (HCC) cells and adeno-associated viral vectors in livers of mice. We assessed proliferation, differentiation, and chromatin compaction in cultured cells, and liver regeneration and tumor formation in mice. Reducing levels of α1-ACT promoted proliferation of HCC cells in vitro. Oncostatin M up-regulated α1-ACT expression and nuclear translocation, which inhibited HCC cell proliferation and activated differentiation of mouse hepatocytes. We identified amino acids required for α1-ACT nuclear localization, and found that α1-ACT inhibits cell-cycle progression and anchorage-independent proliferation of HCC cells. HCC cells that overexpressed α1-ACT formed smaller tumors in mice than HCC cells that did not express the protein. α1-ACT was observed to self-associate and polymerize in the nuclei of cells; nuclear α1-ACT strongly bound chromatin to promote a condensed state that could prevent cell proliferation. α1-ACT localizes to the nuclei of hepatic cells to control chromatin condensation and proliferation. Overexpression of α1-ACT slows the growth of HCC xenograft tumors in nude mice.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti--actina antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anticuerpo anti-trimetil-histona H3 (Lys27), Upstate®, from rabbit
Sigma-Aldrich
Anti-SERPINA3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution