Merck
CD100 and plexins B2 and B1 mediate monocyte-endothelial cell adhesion and might take part in atherogenesis.
Molecular immunology (2015-08-16)
Maria Carolina A Luque, Paulo S Gutierrez, Victor Debbas, Jorge Kalil, Beatriz S Stolf
RESUMEN
Leukocyte migration is essential for the function of the immune system. Their recruitment from the vessels to the tissues involves sequential molecular interactions between leukocytes and endothelial cells (ECs). Many adhesion molecules involved in this process have already been described. However, additional molecules may be important in this interaction, and here we explore the potential role for CD100 and plexins in monocyte-EC binding. CD100 was shown to be involved in platelet-endothelial cell interaction, an important step in atherogenesis and thrombus formation. In a recent work we have described CD100 expression in monocytes and in macrophages and foam cells of human atherosclerotic plaques. In the present work, we have identified plexin B2 as a putative CD100 receptor in these cells. We have detected CD100 expression in the endothelium as well as in in vitro cultured endothelial cells. Blocking of CD100, plexin B1 and/or B2 in adhesion experiments have shown that both CD100 and plexins act as adhesion molecules involved in monocyte-endothelial cell binding. This effect may be mediated by CD100 expressed in both cell types, probably coupled to the receptors endothelial plexin B1 and monocytic plexin B2. These results can bring new insights about a possible biological activity of CD100 in monocyte adhesion and atherosclerosis, as well as a future candidate for targeting therapeutics.