Merck
Low prevalence of heparin-induced thrombocytopenia after cardiac surgery in Thai patients.
Thrombosis research (2014-09-11)
Kochawan Boonyawat, Pantep Angchaisuksiri, Katcharin Aryurachai, Suchart Chaiyaroj, Zohra Ahmadi, Beng Hock Chong
RESUMEN
Heparin induced-thrombocytopenia (HIT) has been well recognized in Western countries. However, there are no data in the Thai population. We therefore investigated the prevalence of anti-platelet factor 4 (PF4)/heparin antibodies, HIT, and its thrombotic complications in Thai patients undergoing cardiac surgery using unfractionated heparin. Seventy-three consecutive patients were prospectively enrolled in this study. Blood samples before operation and week 1, week 2, and week 3 after operation were collected from each patient for HIT antibody screening by enzyme-linked immunosorbent assay using IgG antibody specific to the PF4/heparin complex. Positive samples were further analyzed by (14)C-serotonin release assay. Complete blood count was performed daily during the first week, then weekly for 3 weeks. No patient had detectable anti-PF4/heparin antibodies at baseline. Five patients sero-converted during the course of the study for anti-PF4/heparin IgG: 3 (4.1%) at week 1, 4 (5.5%) at week 2, and 5 (6.8%) at week 3 after surgery. However, none of these patients had anti-PF4/heparin antibodies that resulted in (14)C-serotonin release to be considered clinically significant antibodies. Post-operative thrombocytopenia after the operation was found in 35 patients (47.9%), but was not considered to be caused by HIT. Thromboembolic events occurred in 3 patients (4.1%) during follow up; however, none of these patients had positive PF4/heparin antibody tests. Our study represents the first study to examine Thai patients exposed to heparin in the context of cardiac surgery. We found a lower prevalence of positive anti-PF4/heparin antibodies and clinical HIT than previously published studies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Human Platelet Factor-4 / CXCL4 ELISA Kit, for serum, plasma, cell culture supernatant and urine