Merck
Immunophenotypic analysis of myeloperoxidase-negative leukemia cutis and blastic plasmacytoid dendritic cell neoplasm.
American journal of clinical pathology (2012-02-18)
Danielle M P Cronin, Tracy I George, Kaaren K Reichard, Uma N Sundram
RESUMEN
Myeloid leukemia cutis (LC) and blastic plasmacytoid dendritic cell neoplasm (BPDCN) are morphologically indistinguishable malignancies that frequently manifest in the skin. Separating myeloperoxidase-negative LC from BPDCN may be particularly challenging. We identified a panel of immunohistochemical stains to distinguish myeloid LC (23 cases) from BPDCN (12 cases): myeloperoxidase, which stained 7 cases (30%) of LC and 0 cases (0%) of BPDCN; CD56, which stained 12 cases (52%) of LC and all 12 cases (100%) of BPDCN; CD4, which stained 2 cases (9%) of LC and all 12 cases (100%) of BPDCN; CD123, which stained 4 cases (17%) of LC and 10 cases (83%) of BPDCN; and Tcl-1, which stained 2 cases (9%) of LC and 9 (82%) of 11 cases of BPDCN. It is interesting that CD33 was not helpful; it stained 18 (78%) cases of LC and 11 cases (92%) of BPDCN. Our results indicate that a panel that includes CD4, CD56, CD123, and Tcl-1 can appropriately distinguish between these 2 entities.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
CD123 (6H6) Mouse Monoclonal Antibody