Merck
Isolation and characterization of a MAGE gene family in the Xp21.3 region.
Proceedings of the National Academy of Sciences of the United States of America (1995-05-23)
F Muscatelli, A P Walker, E De Plaen, A N Stafford, A P Monaco
RESUMEN
A human gene with strong homology to the MAGE gene family located in Xq27-qter has been isolated by using exon-trapping of cosmids in the Xp21.3 region. We have mapped and sequenced cDNA and genomic clones corresponding to this gene, MAGE-Xp, and shown that the last exon contains the open reading frame and is present in a minimum of five copies in a 30-kb interval. MAGE-Xp is expressed only in testis and, unlike the Xq27-qter MAGE genes, it is not expressed in any of 12 different tumor tissues tested. However, the gene and predicted protein structure are conserved, suggesting a similar function. MAGE-Xp is located in the 160-kb critical interval defined for the locus involved in sex determination within Xp21 and is 50 kb distal to the DAX-1 gene, which is responsible for X-chromosome-linked adrenal hypoplasia congenita.