Merck
Squaramide-catalyzed enantioselective Friedel-Crafts reaction of indoles with imines.
Chemical communications (Cambridge, England) (2010-04-14)
Yong Qian, Gaoyuan Ma, Aifeng Lv, Hai-Liang Zhu, Jing Zhao, Viresh H Rawal
RESUMEN
Chiral squaramides are highly enantioselective catalysts for Friedel-Crafts reaction of indoles with N-tosyl imines, affording 3-indolyl methanamine products in 85-96% yields and 84-96% ees.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
3,4-Dimethoxy-3-cyclobutene-1,2-dione, 99%