Merck
Clinical experiences with trandolapril.
American heart journal (1993-05-01)
C A Gaillard, P W de Leeuw
RESUMEN
In this article the clinical experiences with the new non-sulfhydryl angiotensin-converting enzyme inhibitor trandolapril are reviewed. The drug lowers blood pressure effectively in patients with mild-to-severe hypertension including elderly and obese patients. Distinguishing features of the compound are its long plasma half-life and high degree of lipophilicity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Trandolapril, ≥98% (HPLC), white, powder
Trandolapril, European Pharmacopoeia (EP) Reference Standard