Merck
Transforming growth factor-beta is activated by plasmin and inhibits smooth muscle cell death in human saphenous vein.
Journal of vascular research (2005-05-05)
Sarah J George, Jason L Johnson, Melanie A Smith, Gianni D Angelini, Christopher L Jackson
RESUMEN
The effect of activation of endogenous transforming growth factor-beta (TGF-beta) on smooth muscle cell apoptosis was assessed in human saphenous vein. Segments of human saphenous vein, obtained at the time of bypass graft surgery, were cultured for 14 days. During this time, smooth muscle cells accumulated in the intima as a result of proliferation and migration, partly counterbalanced by apoptotic cell death. Addition of exogenous TGF-beta(1) had no effect on smooth muscle cell proliferation or apoptosis. However, antibody neutralization of endogenous TGF-beta(1) caused significant increases in smooth muscle cell death in the media and intima without any change in proliferation. A plasmin inhibitor (alpha-N-acetyl-L-lysine methyl ester), a specific urokinase-type plasminogen activator (uPA) inhibitor (amiloride) and an anti-catalytic anti-uPA antibody all caused decreases in the tissue content of active TGF-beta and increases in smooth muscle cell death in the media and intima. These data suggest that the amount of TGF-beta in human saphenous vein is sufficient, when in the active form, to protect smooth muscle cells against apoptosis. Adding exogenous TGF-beta(1) has no beneficial effect, but decreasing the amount of active TGF-beta causes smooth muscle cells to undergo apoptosis. Plasmin, generated by uPA, appears to be an important activator of endogenous latent TGF-beta.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Nα-Acetyl-L-lysine methyl ester hydrochloride, 98%