Merck
Phenol coupling initiated by one-electron oxidation of tyrosine units in peptides and histone.
International journal of radiation biology and related studies in physics, chemistry, and medicine (1983-08-01)
W A Prütz, J Butler, E J Land
RESUMEN
Phenoxyl radicals generated pulse radiolytically by the reaction of N.3 with Gly-Tyr decay biomolecularly (2k = 4.7 X 10(8)M-1 s-1) with efficient formation of 2,2'-dimers, which enolize rapidly (k = 2.7 X 10(4) s-1) to produce the 2,2'-biphenolic product. The build-up of the characteristic 2,2'-biphenol fluorescence (400 nm) and absorption also indicated a delayed (k = 80 s-1) process, probably involving the phenoxyl <-> phenoxy-quinol equilibrium. About 60 per cent of the Gly-Tyr phenoxyls were found to dimerize to the 2,2'-biphenol, and a similarly efficient 2,2'-coupling seems to occur with other tyrosyls, such as Lys-Tyr-Lys and histone. gamma-Radiolysis was applied to estimate relative yields of formation of 2,2'-biphenols under various conditions. Dimerization is almost completely inhibited by cysteine or oxygen, consistent with phenoxyl 'repair' by cysteine or O-.2; disproportionation of O-.2 with SOD prevents repair. The phenol 2,2'-coupling is less efficient for .OH- and inefficient for e-aq-initiation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2,2′-Biphenol, 99%