Merck
  • Vitamin B-12 supplementation of rural Mexican women changes biochemical vitamin B-12 status indicators but does not affect hematology or a bone turnover marker.
Vitamin B-12 supplementation of rural Mexican women changes biochemical vitamin B-12 status indicators but does not affect hematology or a bone turnover marker.
The Journal of nutrition (2012-08-24)
Setareh Shahab-Ferdows, Miriam A Anaya-Loyola, Haydé Vergara-Castañeda, Jorge L Rosado, William R Keyes, John W Newman, Joshua W Miller, Lindsay H Allen
RESUMEN
A high prevalence of low serum vitamin B-12 concentrations has been reported in studies and surveys in Latin America including Mexico, but the functional consequences are unknown. This randomized controlled trial assessed the response to a high-dose vitamin B-12 supplementation of women in rural Querétaro, Mexico. Participants aged 20-59 y were stratified at baseline to deficient, marginal, and adequate status groups (serum vitamin B-12, 75-148, 149-220, and >220 pmol/L, respectively), and each group was randomized to vitamin B-12 treatment (single dose of 1 mg i.m. then 500 μg/d orally for 3 mo, n = 70) or placebo (n = 62). Measures at baseline and 3 mo included: complete blood count, serum vitamin B-12, holotranscobalamin (holoTC), folate, ferritin, C-reactive protein (CRP), bone alkaline phosphatase, and methylmalonic acid (MMA) and plasma total homocysteine (tHcy). At baseline, 11% of the women were vitamin B-12 deficient and 22% had marginal status. HoloTC was low (<35 pmol/L) in 23% and correlated with serum vitamin B-12 (r = 0.7; P < 0.001). Elevated MMA (>271 nmol/L) and tHcy (>12 μmol/L) occurred in 21 and 31%, respectively, and correlated with serum vitamin B-12 (r = -0.28, P < 0.0007 and r = -0.20, P < 0.01, respectively). Supplementation increased serum vitamin B-12 and holoTC and lowered MMA and tHcy, normalizing all values except for elevated tHcy in 21% of the women. Supplementation did not affect hematology or bone-specific alkaline phosphatase. Vitamin B-12 supplementation normalized biochemical indicators of vitamin B-12 status in the treatment group but did not affect the functional outcomes measured.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methylmalonic acid, 99%
Supelco
Methylmalonic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®