Merck
Effect of mouth rinses with fluoride and trimetaphosphate on enamel erosion: an in vitro study.
Caries research (2011-10-05)
M M Manarelli, A E M Vieira, A A Matheus, K T Sassaki, A C B Delbem
RESUMEN
The effect of mouth rinses containing fluoride (100 μg/ml) and sodium trimetaphosphate (TMP) on enamel erosion was evaluated in vitro. Bovine enamel blocks were subjected to erosive challenges 4 times per day for 5 min, followed by treatment with placebo, 225 μg F/ml, 100 μg F/ml, 100 μg F/ml and TMP (0.2, 0.4 and 0.6%) solutions (30 s) and storage in artificial saliva, over a duration of 5 days. TMP groups showed lower enamel wear than fluoride-only and placebo groups (p < 0.05). Addition of TMP at a TMP:NaF molar proportion between 1.24:1 and 3.72:1 to a solution containing 100 μg F/ml presented a greater protective effect under erosive conditions than a solution containing 225 μg F/ml, in the absence of TMP.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Trisodium trimetaphosphate, ≥95%