Merck
Multicentre patch testing with a resol resin based on phenol and formaldehyde.
Contact dermatitis (2011-05-21)
Marléne Isaksson, Annica Inerot, Carola Lidén, Mihaly Matura, Berndt Stenberg, Halvor Möller, Magnus Bruze
RESUMEN
Contact allergy to phenol-formaldehyde resins (PFRs) based on phenol and formaldehyde is not detected by a p-tertiary-butylphenol-formaldehyde resin (PTBP-FR) included in most baseline patch test series. To investigate the rate of contact allergy to PFR-2 (a mixture of monomers and dimers from a resol resin based on phenol and formaldehyde) in a Swedish population, and to investigate associated simultaneous allergic reactions. Five centres representing the Swedish Contact Dermatitis Research Group included PFR-2 in their patch test baseline series for a period of 1.5 years. Of 2504 patients tested, 27 (1.1%) reacted to PFR-2. Of those 27 individuals, 2 had a positive reaction to formaldehyde and 2 to PTBP-FR. Simultaneous allergic reactions were noted to colophonium in 6, to Myroxylon pereirae in 14, and to fragrance mix I in 15. The contact allergy frequency in the tested population (1.1%) merits its inclusion in the Swedish baseline series and possibly also in other baseline series. Simultaneous allergic reactions were noted to colophonium, M. pereirae, and fragrance mix I.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Gum rosin, natural resin
Sigma-Aldrich
Gum rosin, CP