Merck
Thymus-independent class switch recombination is affected by APRIL.
Immunology and cell biology (2008-03-26)
Gijs Hardenberg, Liesbeth van Bostelen, Michael Hahne, Jan Paul Medema
RESUMEN
The tumour necrosis factor (TNF) family member a proliferation-inducing ligand (APRIL) is implicated in various B-cell processes, such as class switch recombination, plasma cell differentiation and plasma cell survival. This was suggested from initial studies analysing B-cell responses in APRIL-deficient and transgenic mice, and mice deficient for the TNF receptors of APRIL, transmembrane activator and CAML interactor (TACI) and B-cell maturation antigen. Here, we present additional evidence for the importance of APRIL in thymus-independent (TI) B-cell responses, using APRIL-deficient and transgenic mice. APRIL-deficient mice show an impaired immunoglobulin A (IgA) response towards TI B-cell antigens, whereas APRIL transgenic mice show exaggerated TI B-cell responses. Moreover, antibody titres to TI antigens were sustained in APRIL transgenic mice for a long time and even increased up to 75 days in the case of IgA against 4-hydroxy-nitrophenacetyl-lipopolysaccharide.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Hydroxy-3-nitrophenylacetic acid, 99%