Merck
Isolation and structures of novel fungal metabolites as chemokine receptor (CCR2) antagonists.
The Journal of antibiotics (2006-02-10)
Kithsiri B Herath, Hiranthi Jayasuriya, John G Ondeyka, Jon D Polishook, Gerald F Bills, Anne W Dombrowski, Angeles Cabello, Pasquale P Vicario, Hans Zweerink, Ziqiang Guan, Sheo B Singh
RESUMEN
The chemokine receptor, CCR2, is predominantly expressed on monocytes/macrophages, and on a subset of memory T cells. It binds to several CC type chemokines of the monocyte chemoattractant protein (MCP) family of which MCP-1 exhibits the highest affinity. CCR2/MCP-1 expression/association in monocyte/macrophage/T cells has been associated with inflammatory processes such as rheumatoid arthritis, multiple sclerosis and atherosclerosis. Neutralization of CCR2 with either a peptide or receptor antagonist results in the prevention of joint swelling in rodent models of arthritis. In this paper, bioassay-guided discovery of CCR2 receptor antagonists derived from natural product extracts are reported. These antagonists belong to two main classes exemplified by bisthiodiketopiperazines and cytochalasins. Six compounds, including emestrin, two new emestrin analogs, and chaetomin represent the first group of compounds. These compounds inhibited the binding of MCP-1 to CCR2 (CHO membrane) with IC50 values of 0.8 to 9 microM and exhibited good activity in a whole cell assay using MCP-1 and human monocytes with IC50's ranging from 4-9 microM. Cytochalasins A and B represented the second group and inhibited the binding activity with IC50 values of 5 and 188 microM, respectively. This is the first report of natural product antagonists of the CCR2 receptor.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Albumin, Bovine Serum, 30% Aqueous Solution, Stabilizer-Free
Sigma-Aldrich
Chetomin, from Chaetomium cochliodes, ≥98% (HPLC)