Merck
Studies of the influence of different cross-linking reagents on the immune response against a B-epitope.
Bioconjugate chemistry (1992-01-01)
A Delmas, A Brack, Y Trudelle
RESUMEN
We have previously shown that the carrier polytuftsin obtained by polycondensation of tuftsin, a naturally occurring macrophage activator, increases significantly the antibody response against a linked B-epitope. In the present work, we have studied the influence of different cross-linking reagents on the quality of the conjugation and on the immune response, at both B-cell and T-cell levels. We observed that the cross-linking method used for coupling the B-epitope to the carrier influences the immune response. A hypothesis is put forward to explain the differences observed.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
S-Acetylthioglycolic acid N-hydroxysuccinimide ester, ≥95% (TLC), powder