Merck
Membranoproliferative glomerulonephritis with C3NeF and genetic complement dysregulation.
Pediatric nephrology (Berlin, Germany) (2010-12-29)
Valérie Leroy, Véronique Fremeaux-Bacchi, Michel Peuchmaur, Véronique Baudouin, Georges Deschênes, Marie-Alice Macher, Chantal Loirat
RESUMEN
The development of membranoproliferative glomerulonephritis (MPGN) is associated with uncontrolled activation of the complement alternative pathway. This dysregulation is related either to C3 nephritic factor (C3NeF), an auto-antibody directed against the alternative C3 convertase, or to homozygous loss-of-function mutation of the complement regulatory protein factor H. Heterozygous mutations in the genes coding for factor H, or for the other alternative pathway inhibitory proteins factor I and membrane cofactor protein, have recently been identified in a small number of patients with MPGN with exclusive C3 deposits. We report three hypocomplementemic children with dense deposit disease (n=1) or immune-complex-mediated MPGN type I (n=2), associated with both C3NeF activity and heterozygous mutation of factor H or factor I. These observations highlight the possible combination of genetic and acquired defect in complement control in various subtypes of MPGN, a finding that may influence the treatment strategy in some patients.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Complement factor I from human plasma, >90% (SDS-PAGE)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Referencia (SKU) del distribuidorTamaño de envaseDisponibilidadPrecio
2R-0839-500MG500 mg
Se envía en 21 días desde el socio de Aldrich
EUR 628.30
2R-0839-1G1 g
Se envía en 21 días desde el socio de Aldrich
EUR 1,205.80
2R-0839-2G2 g
Se envía en 21 días desde el socio de Aldrich
EUR 2,188.30
2R-0839-5G5 g
Se envía en 21 días desde el socio de Aldrich
EUR 4,670.80
To order products, please Contacto your local dealer.