Merck
Multidrug resistance reversal effects of aminated thioxanthones and interaction with cytochrome P450 3A4.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques (2012-03-01)
Andreia Palmeira, Emília Sousa, Miguel X Fernandes, Madalena M Pinto, M Helena Vasconcelos
RESUMEN
Aminated thioxanthones have recently been described as dual-acting agents: growth inhibitors of leukemia cell lines and P-glycoprotein (P-gp) inhibitors. To evaluate the selectivity profile of thioxanthones as inhibitors of multidrug resistance (MDR), their interaction with other ABC transporters, which were found to have a strong correlation with multidrug resistance, such as multidrug resistant proteins 1 (MRP1), 2 (MRP2) and 3 (MRP3) and breast cancer resistance protein (BCRP) was also evaluated. The interaction of thioxanthones with cytochrome P450 3A4 (CYP3A4) together with the prediction of their binding conformations and metabolism sites was also investigated. The UIC2 monoclonal antibody-labelling assay was performed using P-gp overexpressing leukemia cells, K562Dox, incubated with eight thioxanthonic derivatives, in order to confirm their P-gp inhibitory activity. A colorimetric-based ATPase assay using membrane vesicles from mammalian cells overexpressing a selected human ABC transporter protein (P-gp, MRP1, MRP2, MRP3, or BCRP) was performed. To verify if some of the thioxanthonic derivatives were substrates or inhibitors of CYP3A4, a luciferin-based luminescence assay was performed. Finally, the in silico prediction of the most probable metabolism sites and docking studies of thioxanthones on CYP3A4 binding site were investigated. Thioxanthones interacted not only with P-gp but also with MRP and BCRP transporters. These compounds also interfere with CYP3A4 activity in vitro, in accordance with the in silico prediction. Thioxanthonic derivatives are multi-target compounds. A better characterization of the interactions of these compounds with classical resistance mechanisms may possibly identify improved treatment applications.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cytochrome P450 human, 3A4 isozyme microsomes, with P450 reductase and cytochrome b5, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)