Merck
Proteomics analysis identifies PEA-15 as an endosomal phosphoprotein that regulates α5β1 integrin endocytosis.
Scientific reports (2021-10-08)
Maisel J Caliva, Won Seok Yang, Shirley Young-Robbins, Ming Zhou, Hana Yoon, Michelle L Matter, Mark L Grimes, Thomas Conrads, Joe William Ramos
RESUMEN
Endosomal trafficking of cell surface receptors is essential to their function. Integrins are transmembrane receptors that integrate adhesion to the extracellular matrix with engagement of the cytoskeleton. Ligated integrins mediate diverse signals that regulate matrix assembly, cell survival, cell morphology, and cell motility. Endosomal trafficking of integrins modulates these signals and contributes to cell motility and is required for cancer cell invasion. The phosphoprotein PEA-15 modulates integrin activation and ERK MAP Kinase signaling. To elucidate novel PEA-15 functions we utilized an unbiased proteomics approach. We identified several binding partners for PEA-15 in the endosome including clathrin and AP-2 as well as integrin β1 and other focal adhesion complex proteins. We confirmed these interactions using proximity ligation analysis, immunofluorescence imaging, pull-down and co-immunoprecipitation. We further found that PEA-15 is enriched in endosomes and was required for efficient endosomal internalization of α5β1 integrin and cellular migration. Importantly, PEA-15 promotion of migration was dependent on PEA-15 phosphorylation at serines 104 and 116. These data support a novel endosomal role for PEA-15 in control of endosomal trafficking of integrins through an association with the β1 integrin and clathrin complexes, and thereby regulation of cell motility.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Rab5 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution